Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYZL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199951.2 | NP_001186880.1 | 2 | 342 | ||
NM_020426.3 | NP_065159.1 | 2 | 456 | |||
GenBank mRNA | AY359018.1 | - | 1 | 260 | ||
AY742214.1 | - | 1 | 232 | |||
BC054481.1 | - | 2 | 302 | 6620264 | ||
BG195727.1 | - | 1 | 310 | |||
DB462957.1 | - | 2 | 445 | |||
GQ472214.1 | - | 2 | 302 | |||
HY024713.1 | - | 2 | 460 |