Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011547.2 | NP_001011547.2 | 14 | 2816 | ||
NM_001135181.1 | NP_001128653.1 | 15 | 2891 | |||
XM_011540924.2 | XP_011539226.1 | 14 | 4497 | |||
XM_011540926.2 | XP_011539228.1 | 13 | 3077 | |||
XM_011540927.1 | XP_011539229.1 | 12 | 2675 | |||
XM_011540928.1 | XP_011539230.1 | 14 | 2695 | |||
GenBank mRNA | AK131200.1 | - | 14 | 2769 | ||
BC025411.1 | - | 5 | 1616 | |||
BX648549.1 | - | 15 | 2845 |