Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001370.1 | NP_001361.1 | 51 | 8585 | ||
XM_006711956.2 | XP_006712019.1 | 51 | 8690 | |||
XM_011532649.2 | XP_011530951.1 | 51 | 8539 | |||
XM_011532650.2 | XP_011530952.1 | 51 | 8540 | |||
XM_011532652.1 | XP_011530954.1 | 50 | 8459 | |||
XM_011532653.1 | XP_011530955.1 | 50 | 8438 | |||
XM_011532654.1 | XP_011530956.1 | 50 | 8318 | |||
XM_011532655.2 | XP_011530957.1 | 47 | 7607 | |||
XM_011532657.1 | XP_011530959.1 | 45 | 7346 | |||
XM_017003521.1 | XP_016859010.1 | 49 | 8312 | |||
XM_017003522.1 | XP_016859011.1 | 46 | 8411 | |||
XM_017003523.1 | XP_016859012.1 | 46 | 7578 | |||
GenBank mRNA | AB051484.2 | - | 15 | 2520 | ||
AB290165.1 | - | 17 | 2724 | |||
BC117259.1 | - | 4 | 717 |