Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284334.1 | NP_001271263.1 | 6 | 551 | ||
NM_014509.4 | NP_055324.2 | 8 | 743 | |||
NR_104300.1 | 8 | 759 | ||||
NR_104301.1 | 8 | 743 | ||||
XM_017028738.1 | XP_016884227.1 | 7 | 690 | |||
XM_017028739.1 | XP_016884228.1 | 7 | 688 | |||
GenBank mRNA | AK294584.1 | - | 8 | 703 | ||
AK295223.1 | - | 7 | 607 | |||
AL450314.1 | - | 8 | 686 | |||
AL589866.1 | - | 1, 2 | 44 | |||
AL590118.1 | - | 6 | 392 | |||
AL590120.1 | - | 8 | 584 | |||
BC119650.2 | - | 8 | 627 | 40115836 | ||
BC119651.1 | - | 8 | 627 | 40115837 | ||
BC127635.1 | - | 8 | 628 | 40115838 | ||
BC127636.1 | - | 8 | 628 | 40115841 | ||
CR456455.1 | - | 1, 2 | 50 |