Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010875.3 | NP_001010875.1 | 4, 5 | 517 | ||
NM_001286806.1 | NP_001273735.1 | 3, 4 | 398 | |||
NM_001286807.1 | NP_001273736.1 | 4, 5 | 575 | |||
XM_005266321.2 | XP_005266378.1 | 4, 5 | 443 | |||
XM_005266322.1 | XP_005266379.1 | 4, 5 | 443 | |||
XM_006719793.3 | XP_006719856.1 | 4, 5 | 689 | |||
XM_006719794.1 | XP_006719857.1 | 4, 5 | 567 | |||
XM_011535026.1 | XP_011533328.1 | 3, 4 | 294 | |||
XM_011535027.2 | XP_011533329.1 | 3, 4 | 294 | |||
XM_017020523.1 | XP_016876012.1 | 4, 5 | 514 | |||
XM_017020524.1 | XP_016876013.1 | 4, 5 | 567 | |||
GenBank mRNA | AK094133.1 | - | 4, 5 | 444 | ||
AK095465.1 | - | 3, 4 | 297 | |||
AK302967.1 | - | 3, 4 | 325 | |||
BC132739.1 | - | 4, 5 | 440 | 40146761 | ||
BC136760.1 | - | 4, 5 | 440 | 9020750 | ||
BC144401.1 | - | 4, 5 | 498 | |||
DA184634.1 | - | 2, 3 | 251 | |||
DC322809.1 | - | 4, 5 | 474 |