Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206796.2 | NP_001193725.1 | 4 | 1053 | ||
NM_001206799.1 | NP_001193728.1 | 3 | 383 | |||
NM_001316318.1 | NP_001303247.1 | 3 | 936 | |||
NM_002654.5 | NP_002645.3 | 3 | 716 | |||
NM_182470.3 | NP_872270.1 | 3 | 932 | |||
NM_182471.3 | NP_872271.1 | 3 | 716 | |||
XM_005254443.1 | XP_005254500.1 | 4 | 440 | |||
XM_005254445.4 | XP_005254502.1 | 3 | 277 | |||
XM_006720570.1 | XP_006720633.1 | 4 | 537 | |||
XM_011521670.2 | XP_011519972.1 | 3 | 502 | |||
XM_017022313.1 | XP_016877802.1 | 3 | 277 | |||
GenBank mRNA | AK092369.1 | - | 3 | 466 | ||
AK222927.1 | - | 3 | 282 | |||
AK294315.1 | - | 4 | 542 | |||
AK299413.1 | - | 3 | 280 | |||
AK307855.1 | - | 4 | 283 | |||
AK309781.1 | - | 3 | 385 | |||
AK312253.1 | - | 3 | 282 | |||
BC000481.2 | - | 3 | 298 | 2964687 | ||
BC007640.1 | - | 3 | 282 | 3533876 | ||
BC007952.2 | - | 3 | 294 | 4299213 | ||
BC012811.2 | - | 3 | 294 | 2958817 | ||
BC023592.2 | - | 3 | 244 | |||
BC035198.1 | - | 3 | 248 | 3859987 | ||
BC096823.1 | - | 3 | 483 | |||
DA382029.1 | - | 3 | 523 | |||
DC326232.1 | - | 3 | 508 | |||
DC376054.1 | - | 3 | 478 | |||
EU794631.1 | - | 3 | 435 | |||
M23725.1 | - | 3 | 303 | |||
M26252.1 | - | 3 | 283 | |||
Z36831.1 | - | 3 | 321 |