Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322854.1 | NP_001309783.1 | 5 | 919 | ||
NM_001322855.1 | NP_001309784.1 | 6 | 1061 | |||
NM_001322856.1 | NP_001309785.1 | 6 | 1018 | |||
NM_001322857.1 | NP_001309786.1 | 5 | 839 | |||
NM_001322858.1 | NP_001309787.1 | 6 | 936 | |||
NM_001322859.1 | NP_001309788.1 | 3 | 545 | |||
NM_001322861.1 | NP_001309790.1 | 2 | 195 | |||
NM_001322862.1 | NP_001309791.1 | 6 | 896 | |||
NM_001322863.1 | NP_001309792.1 | 5 | 868 | |||
NM_001322864.1 | NP_001309793.1 | 4 | 436 | |||
NM_001322865.1 | NP_001309794.1 | 5 | 786 | |||
NM_001322866.1 | NP_001309795.1 | 5 | 759 | |||
NM_001322868.1 | NP_001309797.1 | 5 | 786 | |||
NM_020448.4 | NP_065181.1 | 5 | 781 | |||
NR_136506.1 | 5 | 786 | ||||
NR_136508.1 | 5 | 786 | ||||
XM_011541808.2 | XP_011540110.1 | 4 | 696 | |||
XM_017001868.1 | XP_016857357.1 | 4 | 431 | |||
XM_017001869.1 | XP_016857358.1 | 5 | 766 | |||
XM_017001870.1 | XP_016857359.1 | 6 | 1031 | |||
GenBank mRNA | AK312433.1 | - | 5 | 735 | ||
BC001265.1 | - | 5 | 723 | 3452234 | ||
BC063583.1 | - | 5 | 711 | 4386115 | ||
BX640883.1 | - | 6 | 843 |