Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APITD1-CORT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243768.1 | NP_001230697.1 | 3 | 737 | ||
NM_001270517.1 | NP_001257446.1 | 4 | 673 | |||
NM_198544.3 | NP_940946.1 | 4 | 673 | |||
NR_037187.1 | 2 | 703 | ||||
GenBank mRNA | AF521016.1 | - | 4 | 673 | ||
CF127457.1 | - | 3 | 678 | |||
CF128105.1 | - | 3 | 677 | |||
HY054077.1 | - | 4 | 343 |
Gene Symbol: APITD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199294.2 | NP_954988.1 | 4 | 673 | ||
NR_036462.1 | 4 | 861 | ||||
GenBank mRNA | BC029430.1 | - | 4 | 332 | 4051739 | |
BG213105.1 | - | 1 | 73 | |||
BU902753.1 | - | 3 | 173 |