Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278661.1 | NP_001265590.1 | 5 | 2856 | ||
NM_001278662.1 | NP_001265591.1 | 5 | 2836 | |||
NM_001278663.1 | NP_001265592.1 | 3 | 2633 | |||
NM_001278664.1 | NP_001265593.1 | 3 | 2594 | |||
NM_001278677.1 | NP_001265606.1 | 6 | 2928 | |||
NM_001278678.1 | NP_001265607.1 | 2 | 2498 | |||
NM_203282.3 | NP_975011.3 | 4 | 2721 | |||
XM_011528443.1 | XP_011526745.1 | 6 | 2989 | |||
XM_011528444.1 | XP_011526746.1 | 7 | 3009 | |||
XM_011528448.2 | XP_011526750.1 | 4 | 2668 | |||
XM_017027514.1 | XP_016883003.1 | 6 | 2919 | |||
XM_017027515.1 | XP_016883004.1 | 8 | 3346 | |||
XM_017027516.1 | XP_016883005.1 | 4 | 5393 | |||
XM_017027517.1 | XP_016883006.1 | 5 | 3222 | |||
XM_017027518.1 | XP_016883007.1 | 4 | 2718 | |||
XM_017027519.1 | XP_016883008.1 | 5 | 2997 | |||
GenBank mRNA | AK303431.1 | - | 3 | 2567 | ||
BC043147.1 | - | 4 | 2693 | 5296940 | ||
CN395850.1 | - | 1 | 212 |