Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004340.3 | NP_001004340.2 | 2, 3 | 329 | ||
NM_001017986.3 | NP_001017986.1 | 4, 5 | 605 | |||
XM_017000661.1 | XP_016856150.1 | 4, 5 | 608 | |||
XM_017000662.1 | XP_016856151.1 | 3, 4 | 350 | |||
GenBank mRNA | CD677904.1 | - | 1, 2 | 143 | ||
L03419.1 | - | 4, 5 | 555 | |||
L03420.1 | - | 2, 3 | 279 |