Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020485.4 | NP_065231.3 | 10 | 1471 | ||
NM_138616.3 | NP_619522.3 | 7 | 1018 | |||
NM_138617.3 | NP_619523.3 | 7 | 1076 | |||
NM_138618.3 | NP_619524.3 | 9 | 1337 | |||
XM_005245957.3 | XP_005246014.1 | 8 | 1782 | |||
XM_011541888.2 | XP_011540190.1 | 11 | 1971 | |||
XM_011541889.2 | XP_011540191.1 | 10 | 1486 | |||
XM_011541890.2 | XP_011540192.1 | 9 | 1852 | |||
XM_011541891.2 | XP_011540193.1 | 11 | 1588 | |||
XM_017002014.1 | XP_016857503.1 | 8 | 1731 | |||
GenBank mRNA | AF056965.1 | - | 10 | 1383 | ||
BC139905.1 | - | 10 | 1513 | 40147407 | ||
JX420291.1 | - | 10 | 1385 | |||
JX420292.1 | - | 10 | 1385 | |||
JX420293.1 | - | 10 | 1385 | |||
JX420294.1 | - | 10 | 1385 | |||
JX420295.1 | - | 10 | 1385 | |||
JX420296.1 | - | 10 | 1385 | |||
JX420297.1 | - | 10 | 1385 | |||
JX420298.1 | - | 10 | 1385 | |||
JX420299.1 | - | 10 | 1385 | |||
JX420300.1 | - | 10 | 1385 | |||
JX420301.1 | - | 10 | 1385 | |||
JX420302.1 | - | 10 | 1385 | |||
X54534.1 | - | 10 | 1434 | |||
X63095.1 | - | 7 | 998 | |||
X63096.1 | - | 7 | 938 |