Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBXIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317734.1 | NP_001304663.1 | 2 | 136 | ||
NM_020524.3 | NP_065385.2 | 3 | 225 | |||
GenBank mRNA | AF221521.1 | - | 3 | 174 | ||
AK022497.1 | - | 2 | 95 | |||
AK023219.1 | - | 3 | 166 | |||
AK300544.1 | - | 3 | 189 | |||
AK310388.1 | - | 4 | 654 | |||
AK315551.1 | - | 3 | 206 | |||
BC016852.1 | - | 3 | 153 | 3900461 | ||
HY108652.1 | - | 3 | 225 |