Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020546.2 | NP_065433.2 | 2, 3 | 483 | ||
XM_011513942.2 | XP_011512244.1 | 2, 3 | 430 | |||
GenBank mRNA | AF410885.1 | - | 2, 3 | 442 | ||
AK127036.1 | - | 2, 3 | 431 | |||
AK299775.1 | - | 1, 2 | 72 | |||
AK304041.1 | - | 2, 3 | 318 | |||
BC111743.1 | - | 2, 3 | 421 | 40038765 | ||
BE677757.1 | - | 2, 3 | 481 |