Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286095.1 | NP_001273024.1 | 2 | 139 | ||
NM_020644.2 | NP_065695.1 | 2 | 582 | |||
GenBank mRNA | AB097017.1 | - | 2 | 207 | ||
AK074677.1 | - | 2 | 200 | |||
AK300712.1 | - | 2 | 62 | |||
AK313065.1 | - | 2 | 212 | |||
AY359069.1 | - | 2 | 225 | |||
BC040124.1 | - | 2 | 264 | 6053804 | ||
DA563443.1 | - | 2 | 549 | |||
DC314550.1 | - | 2 | 139 |