Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020649.2 | NP_065700.1 | 1, 2 | 196 | ||
GenBank mRNA | AF174482.1 | - | 1, 2 | 171 | ||
AF266479.2 | - | 1, 2 | 127 | |||
AK074560.1 | - | 1, 2 | 196 | |||
BC008937.2 | - | 1, 2 | 93 | 2821280 | ||
BC009376.2 | - | 1, 2 | 149 | 4121509 | ||
BC019289.1 | - | 1, 2 | 93 | 2821280 |