Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DECR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020664.3 | NP_065715.1 | 4 | 420 | ||
GenBank mRNA | AJ293009.1 | - | 4 | 335 | ||
AK055839.1 | - | 5 | 1066 | |||
AK128012.1 | - | 5 | 538 | |||
AK225961.1 | - | 4 | 589 | |||
BC010740.1 | - | 4 | 406 | 3877040 | ||
BC011968.1 | - | 4 | 325 | 3866503 |