Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020710.2 | NP_065761.1 | 2 | 720 | ||
GenBank mRNA | AB033011.1 | - | 2 | 152 | ||
BC031301.1 | - | 2 | 720 | 4330114 |