Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASKIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020753.4 | NP_065804.2 | 2 | 670 | ||
GenBank mRNA | AF451976.1 | - | 2 | 523 | ||
BC066643.1 | - | 2 | 633 | 5576059 | ||
DA336467.1 | - | 2 | 561 |