Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006657.1 | NP_001006658.1 | 2 | 188 | ||
NM_020779.3 | NP_065830.2 | 2 | 188 | |||
GenBank mRNA | AB037757.2 | - | 2 | 196 | ||
AK122917.1 | - | 2 | 113 | |||
AK315094.1 | - | 2 | 113 | |||
BC036659.2 | - | 2 | 187 | 5266940 |