Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020443.4 | NP_065176.3 | 3 | 1561 | ||
XM_011510097.1 | XP_011508399.1 | 3 | 1240 | |||
XM_011510098.1 | XP_011508400.1 | 3 | 1240 | |||
XM_011510099.1 | XP_011508401.1 | 3 | 1240 | |||
XM_011510100.1 | XP_011508402.1 | 3 | 1240 | |||
XM_011510101.1 | XP_011508403.1 | 3 | 1240 | |||
XM_011510102.1 | XP_011508404.1 | 3 | 1240 | |||
XM_017002751.1 | XP_016858240.1 | 3 | 1240 | |||
XM_017002753.1 | XP_016858242.1 | 3 | 1240 | |||
XM_017002754.1 | XP_016858243.1 | 3 | 1240 | |||
GenBank mRNA | AB032977.3 | - | 3 | 894 | ||
AJ488101.1 | - | 3 | 1092 |