Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020474.3 | NP_065207.2 | 2 | 312 | ||
XM_005258239.3 | XP_005258296.1 | 3 | 380 | |||
XM_017025692.1 | XP_016881181.1 | 3 | 559 | |||
GenBank mRNA | AK293062.1 | - | 4 | 454 | ||
BC038440.1 | - | 3 | 377 | 4478643 | ||
BC047746.1 | - | 2 | 307 | |||
DN996147.1 | - | 2 | 330 | |||
U41514.1 | - | 2 | 249 | |||
X85018.1 | - | 2 | 218 |