Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DBI SIRNA Assays ›
Gene Symbol:
DBI
Gene Name:
diazepam binding inhibitor, acyl-CoA binding protein
Gene Aliases:
ACBD1, ACBP, CCK-RP, EP
Chromosome Location:
Chr.2: 119366924 - 119372551 on Build GRCh38
UniGene ID:
Hs.78888
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28619
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DBI
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079862.2
NP_001073331.1
3, 4
326
NM_001079863.1
NP_001073332.1
3, 4
446
NM_001178017.1
NP_001171488.1
3, 4
465
NM_001178041.2
NP_001171512.1
4, 5
452
NM_001178042.2
NP_001171513.1
4, 5
407
NM_001178043.2
NP_001171514.1
3, 4
356
NM_001282633.1
NP_001269562.1
4, 5
442
NM_001282634.1
NP_001269563.1
4, 5
412
NM_001282635.1
NP_001269564.1
4, 5
442
NM_001282636.1
NP_001269565.1
3, 4
356
NM_020548.7
NP_065438.1
4, 5
437
NR_104221.1
3, 4
605
GenBank mRNA
AM000001.1
-
3, 4
443
BC006466.1
-
3, 4
241
BC062996.1
-
4, 5
406
5168729
BM768076.1
-
3, 4
273
BP294452.1
-
3, 4
268
BP318721.1
-
2, 3
130
BU599304.1
-
3, 4
329
CR456956.1
-
2, 3
198
FM213122.1
-
3, 4
303
FM213123.1
-
4, 5
389
FM213124.1
-
4, 5
384
FM213125.1
-
4, 5
359
FM213126.1
-
4, 5
354
FM213127.1
-
3, 4
465
FM213128.1
-
4, 5
399
FM213129.1
-
3, 4
356
FM213130.1
-
3, 4
605
FM213131.1
-
4, 5
442
FM213132.1
-
4, 5
334
M14200.1
-
3, 4
268
M15887.1
-
3, 4
255
Back To Top
More Information
Panther Classification:
Molecular Function -
transfer/carrier protein
Gene Ontology Categories:
Function(s)
Process(es)
transport
protein palmitoylation
phosphatidylcholine acyl-chain remodeling
lipid binding
benzodiazepine receptor binding
long-chain fatty acyl-CoA binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline