Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTYH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005367.2 | NP_001005367.1 | 10 | 1183 | ||
NM_001201461.1 | NP_001188390.1 | 10 | 1183 | |||
NM_020659.3 | NP_065710.1 | 10 | 1183 | |||
XM_017027025.1 | XP_016882514.1 | 10 | 1027 | |||
XM_017027026.1 | XP_016882515.1 | 10 | 1027 | |||
GenBank mRNA | AB162930.1 | - | 10 | 1066 | ||
AB188496.1 | - | 10 | 1183 | |||
AF177909.1 | - | 10 | 1097 | |||
AK126690.1 | - | 9 | 1094 | |||
AK289468.1 | - | 10 | 1146 | |||
AK293468.1 | - | 11 | 1214 | |||
BC011347.1 | - | 10 | 1088 | 4215136 | ||
BC019358.1 | - | 7 | 516 | 3960435 |