Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008942.1 | NP_001008942.1 | 1 | 183 | ||
GenBank mRNA | AY510340.1 | - | 1 | 183 |
Gene Symbol: Vom1r39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167551.1 | NP_001161023.1 | 1 | 183 |