Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012124.1 | NP_001012124.1 | 1 | 1143 | ||
GenBank mRNA | BC087592.1 | - | 1 | 1143 | 7309941 |