Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfxank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013136.1 | NP_001013154.1 | 5 | 800 | ||
XM_006252898.2 | XP_006252960.1 | 6 | 674 | |||
XM_006252899.2 | XP_006252961.1 | 6 | 711 | |||
XM_006252900.2 | XP_006252962.1 | 5 | 715 | |||
GenBank mRNA | BC087111.1 | - | 5 | 800 | 7193515 | |
FQ231410.1 | - | 5 | 609 |