Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006195.5 | NP_006186.1 | 1, 2 | 313 | ||
NR_024122.1 | 1, 2 | 313 | ||||
NR_024123.1 | 1, 2 | 313 | ||||
XM_006717130.1 | XP_006717193.1 | 1, 2 | 279 | |||
XM_006717131.1 | XP_006717194.1 | 1, 2 | 279 | |||
XM_006717132.1 | XP_006717195.1 | 1, 2 | 279 | |||
XM_006717133.1 | XP_006717196.1 | 1, 2 | 182 | |||
GenBank mRNA | AF355141.1 | - | 1, 2 | 197 | ||
AF355142.1 | - | 1, 2 | 197 | |||
AI479638.1 | - | 277 | ||||
AK289376.1 | - | 1, 2 | 182 | |||
AK297093.1 | - | 1, 2 | 182 | |||
AK312717.1 | - | 1, 2 | 242 | |||
BC094883.1 | - | 1, 2 | 264 | 5212090 | ||
X59841.1 | - | 1, 2 | 309 |