Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020695.3 | NP_065746.3 | 2 | 1219 | ||
XM_011528144.1 | XP_011526446.1 | 2 | 1254 | |||
XM_011528146.2 | XP_011526448.1 | 2 | 1518 | |||
XM_017027028.1 | XP_016882517.1 | 2 | 1254 | |||
XM_017027029.1 | XP_016882518.1 | 2 | 1254 | |||
XM_017027030.1 | XP_016882519.1 | 2 | 1442 | |||
GenBank mRNA | AB032964.1 | - | 2 | 1207 | ||
BC016923.1 | - | 3 | 1373 | |||
BC032244.1 | - | 2 | 1229 | 4398472 |