Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam110a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014050.2 | NP_001014072.2 | 2 | 1009 | ||
NM_001244768.1 | NP_001231697.1 | 2 | 890 | |||
NM_001244769.1 | NP_001231698.1 | 2 | 962 | |||
NM_001244770.1 | NP_001231699.1 | 2 | 809 | |||
XM_006235231.2 | XP_006235293.1 | 2 | 1262 | |||
XM_006235234.2 | XP_006235296.1 | 3 | 997 | |||
GenBank mRNA | BC079251.1 | - | 2 | 1009 | 7133086 | |
CB766675.1 | - | 1 | 122 | |||
CB806736.1 | - | 1 | 299 | |||
DY318143.1 | - | 1 | 690 | |||
JF912497.1 | - | 1 | 620 |