Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ510151.1 | - | 1 | 154 | ||
BC087057.1 | - | 598 | ||||
BC087137.1 | - | 643 | ||||
BC088269.1 | - | 572 | ||||
BC089840.1 | - | 638 | ||||
BC089851.1 | - | 574 | ||||
BC091243.1 | - | 667 | ||||
BC091247.1 | - | 640 | ||||
BC092592.1 | - | 608 | ||||
BC160870.1 | - | 605 |
Gene Symbol: LOC366772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY158661.1 | - | 615 |