Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024999.1 | NP_001020170.1 | 2 | 157 | ||
XM_008772402.1 | XP_008770624.1 | 1 | 1300 | |||
GenBank mRNA | BC091194.1 | - | 2 | 157 | 7373920 |