Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap1-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025135.1 | NP_001020306.1 | 1 | 324 |
Gene Symbol: Krtap1-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727816.2 | XP_002727862.1 | 1 | 490 | ||
XM_008768205.1 | XP_008766427.1 | 2 | 349 |
Gene Symbol: Krtap1-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001055744.4 | XP_001055744.4 | 1 | 451 | ||
XM_008768206.1 | XP_008766428.1 | 2 | 370 | |||
XM_008768207.1 | XP_008766429.1 | 2 | 340 |
Gene Symbol: LOC100365588 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750925.3 | XP_003750973.1 | 1 | 355 |