Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037346.2 | NP_001032423.1 | 2, 3 | 118 | ||
XM_006228411.1 | XP_006228473.1 | 2, 3 | 85 | |||
GenBank mRNA | BC087641.1 | - | 2, 3 | 90 | 7308408 | |
CB720565.1 | - | 2, 3 | 118 | |||
FQ221309.1 | - | 2, 3 | 100 | |||
FQ221510.1 | - | 2, 3 | 101 | |||
FQ224282.1 | - | 2, 3 | 101 | |||
FQ229780.1 | - | 2, 3 | 101 | |||
X51707.1 | - | 2, 3 | 100 |
Gene Symbol: LOC100910336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750647.3 | XP_003750695.1 | 2, 3 | 173 | ||
XM_008766901.1 | XP_008765123.1 | 2, 3 | 85 |
Gene Symbol: LOC103690015 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728672.4 | XP_002728718.4 | 1 | 89 |
Gene Symbol: Rps19l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765600.1 | XP_008763822.1 | 2 | 74 |