Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037510.1 | NP_001032599.1 | 2 | 85 | ||
GenBank mRNA | AY621342.1 | - | 2 | 85 |