Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037512.1 | NP_001032601.1 | 2 | 134 | ||
GenBank mRNA | AY600147.1 | - | 2 | 169 | ||
AY621352.1 | - | 2 | 134 |