Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult2a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012695.3 | NP_036827.3 | 5, 6 | 779 | ||
GenBank mRNA | FQ209484.1 | - | 5, 6 | 654 | ||
FQ209563.1 | - | 5, 6 | 654 | |||
FQ210641.1 | - | 4, 5 | 527 | |||
FQ218388.1 | - | 5, 6 | 655 | |||
FQ219267.1 | - | 5, 6 | 654 | |||
FQ219345.1 | - | 5, 6 | 654 | |||
M33329.1 | - | 5, 6 | 779 | |||
X63410.1 | - | 5, 6 | 779 |