Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gapdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017008.4 | NP_058704.1 | 2, 3 | 92 | ||
GenBank mRNA | AB017801.1 | - | 2, 3 | 58 | ||
BC059110.1 | - | 2, 3 | 93 | 6919611 | ||
BC087743.1 | - | 2, 3 | 61 | 7321006 | ||
U75401.1 | - | 1, 2 | 42 | |||
X02231.1 | - | 2, 3 | 88 |