Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gapdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017008.4 | NP_058704.1 | 3 | 157 | ||
GenBank mRNA | AB017801.1 | - | 3 | 123 | ||
BC059110.1 | - | 3 | 158 | 6919611 | ||
BC087743.1 | - | 3 | 126 | 7321006 | ||
DQ403053.1 | - | 1 | 13 | |||
M17701.1 | - | 2 | 112 | |||
U75401.1 | - | 2 | 107 | |||
X02231.1 | - | 3 | 153 |