Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019132.1 | NP_062005.1 | 263 | |||
GenBank mRNA | BC061967.1 | - | 225 | |||
BC158589.1 | - | 118 | 9099119 | |||
DQ120475.1 | - | 112 | ||||
DQ120476.1 | - | 112 | ||||
FQ212511.1 | - | 90 | ||||
L10326.1 | - | 1 | 99 | |||
M12673.1 | - | 327 | ||||
M17525.1 | - | 263 |