Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022922.2 | NP_075211.2 | 7 | 1237 | ||
GenBank mRNA | BC061781.1 | - | 7 | 1212 | ||
BC126087.1 | - | 7 | 1237 | 8368624 | ||
FQ214607.1 | - | 7 | 1323 | |||
FQ214880.1 | - | 7 | 1250 | |||
FQ216226.1 | - | 6 | 1175 | |||
FQ224920.1 | - | 7 | 1252 | |||
FQ224970.1 | - | 7 | 1245 | |||
L36250.1 | - | 7 | 1227 |
Gene Symbol: LOC100911515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750654.3 | XP_003750702.2 | 7 | 1393 |