Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022922.2 | NP_075211.2 | 7 | 896 | ||
GenBank mRNA | BC061781.1 | - | 7 | 871 | ||
BC126087.1 | - | 7 | 896 | 8368624 | ||
FQ214607.1 | - | 7 | 982 | |||
FQ214670.1 | - | 7 | 905 | |||
FQ214880.1 | - | 7 | 909 | |||
FQ215213.1 | - | 6 | 811 | |||
FQ215529.1 | - | 7 | 908 | |||
FQ215810.1 | - | 7 | 909 | |||
FQ216226.1 | - | 6 | 834 | |||
FQ216307.1 | - | 7 | 795 | |||
FQ216584.1 | - | 7 | 908 | |||
FQ223237.1 | - | 7 | 910 | |||
FQ224880.1 | - | 7 | 907 | |||
FQ224920.1 | - | 7 | 911 | |||
FQ224970.1 | - | 7 | 904 | |||
L36250.1 | - | 7 | 895 |
Gene Symbol: LOC100911515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750654.3 | XP_003750702.2 | 7 | 1052 |