Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042368.2 | NP_001035827.1 | 3, 4 | 571 | ||
NM_001271775.1 | NP_001258704.1 | 3, 4 | 576 | |||
NM_006266.3 | NP_006257.1 | 3, 4 | 592 | |||
GenBank mRNA | AF027169.1 | - | 3, 4 | 487 | ||
AF295773.1 | - | 3, 4 | 507 | |||
AK056462.1 | - | 2, 3 | 269 | |||
AK127524.1 | - | 3, 4 | 576 |