Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134760.1 | NP_001128232.1 | 9 | 1543 | ||
NM_032619.2 | NP_116008.2 | 10 | 1432 | |||
XM_006228691.2 | XP_006228753.1 | 9 | 1687 | |||
XM_006228692.2 | XP_006228754.1 | 10 | 1576 | |||
GenBank mRNA | AB260892.1 | - | 3 | 409 | ||
AF260436.1 | - | 1 | 96 | |||
BC086392.1 | - | 6 | 657 | |||
BC169048.1 | - | 10 | 1387 |