Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006657.1 | NP_001006658.1 | 2 | 195 | ||
NM_020779.3 | NP_065830.2 | 2 | 195 | |||
GenBank mRNA | AB037757.2 | - | 2 | 203 | ||
AK122917.1 | - | 2 | 120 | |||
AK315094.1 | - | 2 | 120 | |||
BC036659.2 | - | 2 | 194 | 5266940 |