Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grpca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181440.1 | NP_852105.1 | 4 | 755 | ||
GenBank mRNA | M58654.1 | - | 4 | 755 |
Gene Symbol: Grpcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763445.1 | XP_008761667.1 | 679 |