Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207589.3 | NP_997472.2 | 3 | 225 | ||
GenBank mRNA | AY319924.1 | - | 3 | 148 | ||
BC088086.1 | - | 3 | 169 | 7326199 | ||
CF979418.1 | - | 3 | 229 | |||
FQ217552.1 | - | 1 | 83 |
Gene Symbol: LOC100359649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FM123695.1 | - | 23 |