Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146216.2 | NP_001139688.1 | 2 | 508 | ||
NM_006283.2 | NP_006274.2 | 2 | 562 | |||
XM_005273625.4 | XP_005273682.1 | 3 | 676 | |||
XM_005273628.1 | XP_005273685.1 | 2 | 183 | |||
XM_005273629.1 | XP_005273686.1 | 3 | 196 | |||
XM_005273630.1 | XP_005273687.1 | 2 | 204 | |||
XM_011544631.1 | XP_011542933.1 | 2 | 562 | |||
XM_011544632.2 | XP_011542934.1 | 4 | 737 | |||
XM_011544633.1 | XP_011542935.1 | 2 | 562 | |||
XM_011544634.1 | XP_011542936.1 | 2 | 562 | |||
XM_011544635.1 | XP_011542937.1 | 2 | 266 | |||
XM_017013766.1 | XP_016869255.1 | 2 | 183 | |||
XM_017013767.1 | XP_016869256.1 | 3 | 196 | |||
XM_017013768.1 | XP_016869257.1 | 2 | 183 | |||
XM_017013769.1 | XP_016869258.1 | 3 | 196 | |||
XM_017013775.1 | XP_016869264.1 | 2 | 204 | |||
XM_017013776.1 | XP_016869265.1 | 3 | 196 | |||
GenBank mRNA | AF049910.1 | - | 2 | 503 | ||
AK294931.1 | - | 2 | 163 | |||
AK295071.1 | - | 2 | 110 | |||
AK304507.1 | - | 4 | 395 | |||
AK314620.1 | - | 2 | 562 | |||
AY072876.2 | - | 2 | 114 | |||
BC041391.1 | - | 4 | 759 | 5276767 | ||
BX647696.1 | - | 2 | 510 | |||
DA553264.1 | - | 2 | 504 | |||
DB084271.1 | - | 2 | 562 |