Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167738.1 | NP_001161210.1 | 27 | 8576 | ||
NM_020443.4 | NP_065176.3 | 30 | 9916 | |||
XM_006711609.2 | XP_006711672.2 | 28 | 8551 | |||
XM_006711610.2 | XP_006711673.2 | 27 | 8527 | |||
XM_006711611.1 | XP_006711674.1 | 28 | 8667 | |||
XM_011510097.1 | XP_011508399.1 | 29 | 9586 | |||
XM_011510098.1 | XP_011508400.1 | 29 | 9571 | |||
XM_011510099.1 | XP_011508401.1 | 28 | 9562 | |||
XM_011510100.1 | XP_011508402.1 | 29 | 9424 | |||
XM_011510101.1 | XP_011508403.1 | 28 | 9400 | |||
XM_011510102.1 | XP_011508404.1 | 27 | 9391 | |||
XM_017002751.1 | XP_016858240.1 | 28 | 9415 | |||
XM_017002752.1 | XP_016858241.1 | 26 | 8518 | |||
GenBank mRNA | AB032977.3 | - | 30 | 9233 | ||
AY043013.1 | - | 30 | 9915 |