Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001130.2 | NP_001001130.1 | 4 | 1962 | ||
GenBank mRNA | BC055337.1 | - | 4 | 1962 | 6413269 | |
BC055756.1 | - | 4 | 1962 | 6413269 |