Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079684.2 | NP_001073152.1 | 8 | 2430 | ||
NM_001079685.2 | NP_001073153.1 | 8 | 2409 | |||
NM_001287335.1 | NP_001274264.1 | 6 | 2143 | |||
NM_001287336.1 | NP_001274265.1 | 7 | 2322 | |||
NM_020444.4 | NP_065177.2 | 9 | 2517 | |||
NR_109796.1 | 5 | 2011 | ||||
NR_109797.1 | 5 | 1978 | ||||
NR_109798.1 | 6 | 2124 | ||||
NR_109799.1 | 6 | 2086 | ||||
NR_109800.1 | 5 | 1999 | ||||
XM_005265941.1 | XP_005265998.1 | 8 | 2330 | |||
XM_005265945.1 | XP_005266002.1 | 6 | 2005 | |||
XM_005265946.4 | XP_005266003.1 | 8 | 2425 | |||
XM_011534595.1 | XP_011532897.1 | 8 | 2292 | |||
XM_011534596.1 | XP_011532898.1 | 7 | 2241 | |||
XM_017009651.1 | XP_016865140.1 | 7 | 2226 | |||
XM_017009652.1 | XP_016865141.1 | 4 | 1949 | |||
XM_017009653.1 | XP_016865142.1 | 6 | 2098 | |||
GenBank mRNA | AB033017.1 | - | 2 | 1528 | ||
AL390217.1 | - | 8 | 2388 | |||
AL832558.1 | - | 1 | 2537 | |||
BC006316.2 | - | 5 | 1931 | |||
BC010448.1 | - | 9 | 2452 | 4155679 | ||
BC012848.1 | - | 6 | 2021 | |||
BC017097.1 | - | 9 | 2474 | 3529175 | ||
BX648859.1 | - | 9 | 2516 |